Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silicon nitride
Linear Formula:
Si
3
N
4
CAS Number:
12033-89-5
Molecular Weight:
140.28
Products
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Particle Size (µm)
Reference Material Type
Special Grade
certified reference material (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
334103
Silicon nitride,
powder, ≥99.9% trace metals basis
Expand
View Pricing
248622
Silicon nitride,
predominantly β-phase, ≤10micron primary particle size
Expand
View Pricing
636703
Silicon nitride,
nanopowder, <50 nm particle size (spherical), ≥98.5% trace metals basis
Expand
View Pricing
325171
Silicon nitride,
predominantly α-phase, ≤10 micron
Expand
View Pricing
NISTRM8983
Silicon nitride,
NIST
®
RM 8983
Expand
View Pricing