Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silicic acid
Empirical Formula (Hill Notation):
H
2
O
3
Si
CAS Number:
7699-41-4
Molecular Weight:
78.10
EC Number:
231-716-3
Products
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Product Line
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
SIL350
Silicic acid,
BioReagent, suitable for column chromatography, 100-200 mesh (75 - 150 μm)
Expand
View Pricing
SILA200
Silicic acid,
suitable for column chromatography, 60-200 mesh
Expand
View Pricing
SILAW
Silicic acid,
BioReagent, suitable for column chromatography, 200-400 mesh (38 - 75 μm)
Expand
View Pricing