Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silica nanoparticles
Empirical Formula (Hill Notation):
SiO
2
CAS Number:
7631-86-9
Molecular Weight:
60.08
Products
Brand
Sigma-Aldrich (10)
Formula Weight
Physical Form
Quality Segment
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
914843
Silica nanoparticles,
pore size non-porous, 150 nm particle size, 5 % (w/w) in water, dispersion
Expand
View Pricing
914622
Silica nanoparticles,
pore size non-porous, 500 nm particle size, 5 % (w/w) in water, dispersion
Expand
View Pricing
913642
Silica nanoparticles,
pore size non-porous, 150 nm particle size, 5 % (w/w) in ethanol, dispersion
Expand
View Pricing
914126
Silica nanoparticles,
pore size non-porous, 300 nm particle size, 5 % (w/w) in water, dispersion
Expand
View Pricing
914142
Silica nanoparticles,
pore size non-porous, 900 nm particle size, 5 % (w/w) in water, dispersion
Expand
View Pricing
913618
Silica nanoparticles,
pore size non-porous, 500 nm particle size, 5 % (w/w) in ethanol, dispersion
Expand
View Pricing
913898
Silica nanoparticles,
pore size non-porous, 300 nm particle size, 5 % (w/w) in ethanol, dispersion
Expand
View Pricing
913626
Silica nanoparticles,
pore size non-porous, 700 nm particle size, 5 % (w/w) in ethanol, dispersion
Expand
View Pricing
914630
Silica nanoparticles,
pore size non-porous, 900 nm particle size, 5 % (w/w) in ethanol, dispersion
Expand
View Pricing
914886
Silica nanoparticles,
pore size non-porous, 700 nm particle size, 5 % (w/w) in water, dispersion
Expand
View Pricing