Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silica gel 60 (0.040-0.063 mm)
Synonym(s):
Silica, Silicon dioxide
Linear Formula:
SiO
2
CAS Number:
7631-86-9
Molecular Weight:
60.08
Products
Brand
Millipore (1)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Separation Technique
Quality Segment
Application
Particle Size (µm)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1.09385
Silica gel 60 (0.040-0.063 mm),
for column chromatography (230-400 mesh ASTM)
Expand
View Pricing