Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SGE Needles for RN syringes
Products
Product Category
needles (3)
Manufacturer
Length (mm)
OD (mm)
Volume (µL)
Compatibility
Needle Shape
Package Size
Gauge
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
24447
SGE Needles for RN syringes,
volume 25-500 μL, needle size 25 ga, needle L × O.D. 50 mm × 0.50 mm
Expand
View Pricing
24437
SGE Needles for RN syringes,
volume 10 μL, N10-5, needle size 26 ga
Expand
View Pricing
24432
SGE Needles for RN syringes,
volume 1 μL, 1B-7C, needle size 23 ga, needle L × O.D. 70 mm × 0.63 mm
Expand
View Pricing