Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sephacryl
®
Size Exclusion Resin
Synonym(s):
Poly([allyl dextran]-
co
-N,N′-methylenebisacrylamide)
CAS Number:
65546-95-4
Products
Brand
Sigma-Aldrich (3)
Physical Form
Quality Segment
Particle Size (µm)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S300HR
Sephacryl
®
Size Exclusion Resin,
300-HR, MW range 10-1500 kDa (globular proteins), MW range 1-400 kDa (dextrans)
Expand
View Pricing
S400HR
Sephacryl
®
Size Exclusion Resin,
400-HR, MW range 20-8000 kDa (globular proteins)
Expand
View Pricing
S500HR
Sephacryl
®
Size Exclusion Resin,
500-HR, MW range 40-20,000 kDa (dextrans)
Expand
View Pricing