
S-Desoxybrevetoxin B2 solution

Synonym(s):
42-[[(2R)-2-Amino-2-carboxyethyl]thio]-42-deoxo-41,43-dihydro-43-hydroxybrevetoxin B
Empirical Formula (Hill Notation):
C53H79NO16S
CAS Number:
Molecular Weight:
1018.26
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1