![(S)-(+)-2-Phenylpropionic acid 97%](/deepweb/assets/sigmaaldrich/product/structures/214/733/c0b2cde0-d861-4d1b-89a5-ad150bc326a8/640/c0b2cde0-d861-4d1b-89a5-ad150bc326a8.png)
(S)-(+)-2-Phenylpropionic acid
![(S)-(+)-2-Phenylpropionic acid 97%](/deepweb/assets/sigmaaldrich/product/structures/214/733/c0b2cde0-d861-4d1b-89a5-ad150bc326a8/640/c0b2cde0-d861-4d1b-89a5-ad150bc326a8.png)
Synonym(s):
(S)-(+)-Hydratropic acid, (S)-HTA
Linear Formula:
CH3CH(C6H5)CO2H
CAS Number:
Molecular Weight:
150.17
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing