Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Screw cap tubes without caps, conical bottom
Products
Product Category
tubes (7)
Color
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
AXYST200X
Screw cap tubes without caps, conical bottom,
size 2.0 mL, amber, pkg of 8x500caps/cs
Expand
View Pricing
AXYST150
Screw cap tubes without caps, conical bottom,
size 1.5 mL, clear, pkg of 8x500caps/cs
Expand
View Pricing
AXYST050X
Screw cap tubes without caps, conical bottom,
size 0.5 mL, amber, pkg of 8x500caps/cs
Expand
View Pricing
AXYST200
Screw cap tubes without caps, conical bottom,
size 2.0 mL, clear, pkg of 8x500caps/cs
Expand
View Pricing
AXYST050
Screw cap tubes without caps, conical bottom,
size 0.5 mL, clear, pkg of 8x500caps/cs
Expand
View Pricing
AXYST45
Screw cap tubes without caps, conical bottom,
size 0.5 mL, clear, pkg of 8x500caps/cs
Expand
View Pricing
AXYST150XS
Screw cap tubes without caps, conical bottom,
capacity 1.5 mL, amber, sterile
Expand
View Pricing