Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Screw cap, solid top with PTFE liner, pkg 100
Products
Product Category
caps (7)
Brand
Supelco (4)
Closure Type
Color
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
27141
Screw cap, solid top with PTFE liner, pkg 100,
solid green melamine resin cap, F217/PTFE liner, for use with 4 mL vial with 13-425 thread
Expand
View Pricing
27091-U
Screw cap, solid top with PTFE liner, pkg 100,
solid green melamine resin cap, F217/PTFE liner, for use with 2 mL vial (standard opening with 8-425 thread)
Expand
View Pricing
27152
Screw cap, solid top with PTFE liner, pkg 100,
green melamine resin solid cap, F217/PTFE liner, for use with 7 mL vial with 15-425 thread
Expand
View Pricing
27163
Screw cap, solid top with PTFE liner, pkg 100,
green melamine resin solid cap, F217/PTFE liner, for use with 15 mL vial with 18 mm thread
Expand
View Pricing
27186
Screw cap, solid top with PTFE liner, pkg 100,
green melamine resin solid cap, F217/PTFE liner, for use with 20 or 40 mL vial with 24-400 thread
Expand
View Pricing
27174-U
Screw cap, solid top with PTFE liner, pkg 100,
green melamine resin solid cap, F217/PTFE liner, for use with 22 mL vial with 20 mm thread
Expand
View Pricing
28659-U
Screw cap, solid top with PTFE liner, pkg 100,
thread for 10-425
Expand
View Pricing