Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Scienceware
®
safety labeled wash bottles
Products
Product Category
bottles (16)
Brand
Sigma-Aldrich (1)
Manufacturer
Capacity (mL)
Cap
Color
Package Size
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
Z423092
Scienceware
®
safety labeled wash bottles,
Acetone
Expand
View Pricing
Z423149
Scienceware
®
safety labeled wash bottles,
Label your own
Expand
View Pricing
Z423335
Scienceware
®
safety labeled wash bottles,
Water
Expand
View Pricing
Z423106
Scienceware
®
safety labeled wash bottles,
Deionized water
Expand
View Pricing
Z423114
Scienceware
®
safety labeled wash bottles,
Ethanol
Expand
View Pricing
Z423122
Scienceware
®
safety labeled wash bottles,
Isopropanol
Expand
View Pricing
Z423130
Scienceware
®
safety labeled wash bottles,
Methanol
Expand
View Pricing
BAF116460628
Scienceware
®
safety labeled wash bottles,
label, Toluene, wide-mouth red cap, capacity 500 mL (16 oz)
Expand
View Pricing
BAF116460627
Scienceware
®
safety labeled wash bottles,
label, Sodium Hypochlorite (Bleach), wide-mouth yellow cap, capacity 500 mL (16 oz)
Expand
View Pricing
BAF116460620
Scienceware
®
safety labeled wash bottles,
label, Distilled water, wide-mouth blue cap, capacity 500 mL (16 oz)
Expand
View Pricing
BAF116460611
Scienceware
®
safety labeled wash bottles,
label, Methyl Ethyl Ketone, wide-mouth green cap, capacity 500 mL (16 oz)
Expand
View Pricing
BAF116466250
Scienceware
®
safety labeled wash bottles,
label, Dichloromethane, wide-mouth yellow cap, capacity 500 mL (16 oz)
Expand
View Pricing
BAF116460459
Scienceware
®
safety labeled wash bottles,
label, Machine Oil (No Diamond), wide-mouth natural cap, capacity 500 mL (16 oz)
Expand
View Pricing
BAF116460614
Scienceware
®
safety labeled wash bottles,
label, Soap (No Diamond), wide-mouth blue cap, capacity 500 mL (16 oz)
Expand
View Pricing
BAF116460612
Scienceware
®
safety labeled wash bottles,
wide-mouth green cap, capacity 500 mL , can, 16oz
Expand
View Pricing
BAF116460617
Scienceware
®
safety labeled wash bottles,
label, Saline Solution (No Diamond), wide-mouth natural cap, capacity 500 mL (16 oz)
Expand
View Pricing