Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Scienceware
®
biohazard disposal bags
Synonym(s):
Scienceware
®
biohazard disposal bags
Products
Product Category
bags (10)
Brand
Sigma-Aldrich (3)
Color
Manufacturer
Capacity (L)
Color
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
B8151
Scienceware
®
biohazard disposal bags,
W × H × thickness 8 1/2 in. (22 cm) × 11 in. (28 cm) × 0.00125 in., red, pack of 100
Expand
View Pricing
BAF131660001
Scienceware
®
biohazard disposal bags,
W × H × thickness 8 1/2 in. (22 cm) × 11 in. (28 cm) × 0.018 mm, orange-red, pack of 1000
Expand
View Pricing
B8276
Scienceware
®
biohazard disposal bags,
W × H × thickness 25 in. (64 cm) × 35 in. (89 cm) × 0.002 in., red, pack of 200
Expand
View Pricing
B4408
Scienceware
®
biohazard disposal bags,
W × H × thickness 24 in. (61 cm) × 36 in. (91 cm) × 0.00125 in., translucent, pack of 100
Expand
View Pricing
BAF131641923
Scienceware
®
biohazard disposal bags,
width × H × thickness 480 mm × 23 in. (580 mm) × 0.04 mm (1.5 mil), pack of 200 ea, red
Expand
View Pricing
BAF131600009
Scienceware
®
biohazard disposal bags,
width × H × thickness 300 mm (12 in.) × 24 in. (610 mm) × 1.5 mil (0.04 mm), pack of 100 ea, translucent polypropylene
Expand
View Pricing
BAF131641419
Scienceware
®
biohazard disposal bags,
width × H × thickness 14 in. (360 mm) × 19 in. (480 mm) × 0.04 mm (1.5 mil), pack of 200 ea, translucent red polypropylene
Expand
View Pricing
BAF131610009
Scienceware
®
biohazard disposal bags,
width × H × thickness 610 mm × 30 in. (760 mm) × 0.04 mm (1.5 mil), translucent polypropylene, pack of 100 ea
Expand
View Pricing
BAF131643138
Scienceware
®
biohazard disposal bags,
width × H × thickness 790 mm (31 in.) × 38 in. (970 mm) × 0.04 mm (1.5 mil), pack of 200 ea, red
Expand
View Pricing
BAF131643848
Scienceware
®
biohazard disposal bags,
width × H × thickness 970 mm (38 in.) × 48 in. (1220 mm) × 0.04 mm (1.5 mil), pack of 100 ea, red
Expand
View Pricing