Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Schlenk reaction and storage tube
Products
Product Category
tubes (14)
Brand
Sigma-Aldrich (9)
Capacity (mL)
Color
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
Z515981
Schlenk reaction and storage tube,
female joint: ST/NS 14/20, capacity 25 mL
Expand
View Pricing
Z516007
Schlenk reaction and storage tube,
female joint: ST/NS 14/20, capacity 50 mL
Expand
View Pricing
Z516023
Schlenk reaction and storage tube,
female joint: ST/NS 24/40, capacity 100 mL
Expand
View Pricing
Z516082
Schlenk reaction and storage tube,
male joint: ST/NS 14/20, capacity 25 mL
Expand
View Pricing
Z516066
Schlenk reaction and storage tube,
female joint: ST/NS 24/40, capacity 250 mL
Expand
View Pricing
Z516015
Schlenk reaction and storage tube,
female joint: ST/NS 14/20, capacity 100 mL
Expand
View Pricing
Z516104
Schlenk reaction and storage tube,
male joint: ST/NS 14/20, capacity 100 mL
Expand
View Pricing
Z516090
Schlenk reaction and storage tube,
male joint: ST/NS 14/20, capacity 50 mL
Expand
View Pricing
Z516112
Schlenk reaction and storage tube,
male joint: ST/NS 24/40, capacity 100 mL
Expand
View Pricing
Z516155
Schlenk reaction and storage tube,
male joint: ST/NS 29/32, capacity 250 mL
Expand
View Pricing
Z516031
Schlenk reaction and storage tube,
female joint: ST/NS 29/32, capacity 100 mL
Expand
View Pricing
Z516120
Schlenk reaction and storage tube,
male joint: ST/NS 29/32, capacity 100 mL
Expand
View Pricing
Z516058
Schlenk reaction and storage tube,
female joint: ST/NS 14/20, capacity 250 mL
Expand
View Pricing
Z516074
Schlenk reaction and storage tube,
female joint: ST/NS 29/32, capacity 250 mL
Expand
View Pricing