
Schizandrin C

Synonym(s):
Schisandrin C, Schizandrin C, Wuweizisu C
Empirical Formula (Hill Notation):
C22H24O6
CAS Number:
Molecular Weight:
384.42
Products
Formula Weight
Manufacturer
Product Line
Format
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing