Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sarcosine
Synonym(s):
N
-Methylglycine, Sarcosine
Linear Formula:
CH
3
NHCH
2
COOH
CAS Number:
107-97-1
Molecular Weight:
89.09
EC Number:
203-538-6
Products
Product Category
amino acids (3)
Brand
Sigma-Aldrich (4)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
131776
Sarcosine,
98%, suitable for cell culture
Expand
View Pricing
8.07666
Sarcosine,
for peptide synthesis
Expand
View Pricing
S7672
Sarcosine,
≥98%, suitable for HPLC, BioXtra
Expand
View Pricing
84532
Sarcosine,
≥98.0% (T)
Expand
View Pricing