
Saccharin sodium salt hydrate

Synonym(s):
2,3-Dihydro-3-oxobenzisosulfonazole sodium salt, 2-Sulfobenzoic acid imide sodium salt, o-Sulfobenzimide sodium salt, Saccharin soluble
Empirical Formula (Hill Notation):
C7H4NNaO3S · xH2O
Products
Product Category
Formula Weight
Physical Form
Product Line
Purity
Showing 1-2 of 2
Compare
Product Number
Description
Pricing