
Riociguat for system suitability

Synonym(s):
Riociguat, N-[4,6-Diamino-2-[1-[(2-fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridin-3-yl]-5-pyrimidinyl]-N-methylcarbamic acid methyl ester
Empirical Formula (Hill Notation):
C20H19FN8O2
CAS Number:
Molecular Weight:
422.42
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing