
Rifabutin

Synonym(s):
Ansamycin, Ansatipine (Farmitalia), LM-427, Mycobutin (Farmitalia), Rifabutin
Empirical Formula (Hill Notation):
C46H62N4O11
CAS Number:
Molecular Weight:
847.00
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Activity Spectrum
Agency
Format
Research Area
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing