Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Rhodium(III) chloride hydrate
Synonym(s):
Rhodium trichloride hydrate
Linear Formula:
RhCl
3
· xH
2
O
CAS Number:
20765-98-4
Molecular Weight:
209.26 (anhydrous basis)
EC Number:
233-165-4
Products
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Reagent Type
Core
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
206261
Rhodium(III) chloride hydrate,
Rh 38-40 %
Expand
View Pricing
520772
Rhodium(III) chloride hydrate,
crystalline, ≥99.9% trace metals basis
Expand
View Pricing
450286
Rhodium(III) chloride hydrate,
99.95% trace metals basis
Expand
View Pricing