Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Red Fluorescent PLGA nanoparticles
Products
Brand
Sigma-Aldrich (7)
Physical Form
Particle Size (µm)
Polymer Composition
Polymer Type
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
LGFR100
Red Fluorescent PLGA nanoparticles,
100 nm average diameter
Expand
View Pricing
LGFR1000
Red Fluorescent PLGA nanoparticles,
1 μm average diameter
Expand
View Pricing
LGFR200
Red Fluorescent PLGA nanoparticles,
200 nm average diameter
Expand
View Pricing
LGFR50K
Red Fluorescent PLGA nanoparticles,
50 μm average diameter
Expand
View Pricing
LGFR500
Red Fluorescent PLGA nanoparticles,
500 nm average diameter
Expand
View Pricing
LGFR10K
Red Fluorescent PLGA nanoparticles,
10 μm average diameter
Expand
View Pricing
LGFR20K
Red Fluorescent PLGA nanoparticles,
20 μm average diameter
Expand
View Pricing