Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(
R
)-(+)-α-Methylbenzylamine
Synonym(s):
(
R
)-(+)-1-Phenylethylamine
Linear Formula:
C
6
H
5
CH(CH
3
)NH
2
CAS Number:
3886-69-9
Molecular Weight:
121.18
EC Number:
223-423-4
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (3)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
115541
(
R
)-(+)-α-Methylbenzylamine,
98%
Expand
View Pricing
726621
(
R
)-(+)-α-Methylbenzylamine,
ChiPros
®
, produced by BASF, ≥99.0%
Expand
View Pricing
77879
(
R
)-(+)-α-Methylbenzylamine,
for chiral derivatization, LiChropur
™
, ≥99.0%
Expand
View Pricing
77880
(
R
)-(+)-α-Methylbenzylamine,
purum, ≥98.0% (sum of enantiomers, GC)
Expand
View Pricing