
Raltegravir Related Compound E

Synonym(s):
N-{2-[4-(Benzylcarbamoyl)-5-hydroxy-1-methyl-6-oxo-1,6-dihydropyrimidin-2-yl]propan-2-yl}-5-methyl-1,3,4-oxadiazole-2-carboxamide
Empirical Formula (Hill Notation):
C20H22N6O5
CAS Number:
Molecular Weight:
426.43
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing