Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
radiello
™
Passive Sampler Starter Kit
Synonym(s):
radiello
™
Starter Kits
Products
Sustainability
Feature
Manufacturer
Agency
Application
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
RAD130S
radiello
™
Passive Sampler Starter Kit,
BTEX/VOC, CS2 Desorption
Expand
View Pricing
RAD145S
radiello
™
Passive Sampler Starter Kit,
BTEX/VOC, Thermal Desorption
Expand
View Pricing
RAD170S
radiello
™
Passive Sampler Starter Kit,
Hydrogen Sulfide
Expand
View Pricing
RAD141S
radiello
™
Passive Sampler Starter Kit,
1, 3-Butadiene, Thermal Desorption
Expand
View Pricing