
Quinuclidine hydrochloride

Synonym(s):
1-Azabicyclo[2.2.2]octane hydrochloride
Empirical Formula (Hill Notation):
C7H13N · HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing