Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Qiagen
®
Multi Barrier Pouch
Products
Manufacturer
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
WHAWB100037
Qiagen
®
Multi Barrier Pouch,
medium, size , L x W 4.37" x 6.5" (11.1 cm x 16.5 cm), pkg of 100 ea
Expand
View Pricing
WHAWB100036
Qiagen
®
Multi Barrier Pouch,
small, size , L x W 3.75" x 3" (9.5 cm x 7.6 cm), pkg of 100 ea
Expand
View Pricing
WHAWB100092
Qiagen
®
Multi Barrier Pouch,
medium, size , L x W 4" x 4.5" (10.1 cm x 11.4 cm), pack of 100 ea
Expand
View Pricing
WHAWB100024
Qiagen
®
Multi Barrier Pouch,
large, size , L x W 7" x 7.37" (17.8 cm x 18.7 cm), resealable, pkg of 50 ea
Expand
View Pricing