Skip to Content
Merck
Pyridoxal 5′-phosphate hydrate powder, BioReagent, suitable for cell culture

Pyridoxal 5′-phosphate hydrate

Pyridoxal 5′-phosphate hydrate powder, BioReagent, suitable for cell culture
Synonym(s):
3-Hydroxy-2-methyl-5-([phosphonooxy]methyl)-4-pyridinecarboxaldehyde, Codecarboxylase, PLP, Pyridoxal 5-phosphate
Empirical Formula (Hill Notation):
C8H10NO6P · xH2O
CAS Number:
Molecular Weight:
247.14 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-2 of 2