Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Measuring cylinder with spout, class A with white graduations
Products
Product Category
cylinders (9)
Manufacturer
Height (mm)
Color
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
SLW3201/16WC
Pyrex
®
Measuring cylinder with spout, class A with white graduations,
works certified, 1000 mL
Expand
View Pricing
SLW3201/08WC
Pyrex
®
Measuring cylinder with spout, class A with white graduations,
works certified, 50 mL
Expand
View Pricing
SLW3201/10WC
Pyrex
®
Measuring cylinder with spout, class A with white graduations,
works certified, 100 mL
Expand
View Pricing
SLW3201/12WC
Pyrex
®
Measuring cylinder with spout, class A with white graduations,
works certified, 250 mL
Expand
View Pricing
SLW3201/14WC
Pyrex
®
Measuring cylinder with spout, class A with white graduations,
works certified, 500 mL
Expand
View Pricing
SLW3201/06WC
Pyrex
®
Measuring cylinder with spout, class A with white graduations,
works certified, 25 mL
Expand
View Pricing
SLW3201/18WC
Pyrex
®
Measuring cylinder with spout, class A with white graduations,
works certified, 2000 mL
Expand
View Pricing
SLW3201/04WC
Pyrex
®
Measuring cylinder with spout, class A with white graduations,
works certified, 10 mL
Expand
View Pricing
SLW3201/02WC
Pyrex
®
Measuring cylinder with spout, class A with white graduations,
works certified, 5 mL
Expand
View Pricing