Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PYREX
®
Disposable Glass Conical Centrifuge Tubes
Products
Product Category
tubes (4)
Color
Manufacturer
Capacity (mL)
Length (mm)
OD (mm)
Cap
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS9950215
PYREX
®
Disposable Glass Conical Centrifuge Tubes,
without cap, capacity 15 mL
Expand
View Pricing
CLS9950210
PYREX
®
Disposable Glass Conical Centrifuge Tubes,
without cap, capacity 10 mL
Expand
View Pricing
CLS9950250
PYREX
®
Disposable Glass Conical Centrifuge Tubes,
without cap, capacity 50 mL
Expand
View Pricing
CLS995025
PYREX
®
Disposable Glass Conical Centrifuge Tubes,
without cap, capacity 5 mL
Expand
View Pricing