Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Putrescine dihydrochloride
Synonym(s):
1,4-Butanediamine dihydrochloride, 1,4-Diaminobutane dihydrochloride, Tetramethylenediamine dihydrochloride
Linear Formula:
NH
2
(CH
2
)
4
NH
2
· 2HCl
CAS Number:
333-93-7
Molecular Weight:
161.07
EC Number:
206-375-9
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (3)
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Markush Class
Markush Group
Sterilization Method
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
P5780
Putrescine dihydrochloride,
powder, BioReagent, suitable for cell culture
Expand
View Pricing
P7505
Putrescine dihydrochloride,
≥98% (TLC), powder
Expand
View Pricing
P6024
Putrescine dihydrochloride,
γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Expand
View Pricing