Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Puromycin dihydrochloride from
Streptomyces alboniger
Synonym(s):
3′-[α-Amino-
p
-methoxyhydrocinnamamido]-3′-deoxy-N,N-dimethyladenosine dihydrochloride
Empirical Formula (Hill Notation):
C
22
H
29
N
7
O
5
· 2HCl
CAS Number:
58-58-2
Molecular Weight:
544.43
EC Number:
200-387-8
Products
Product Category
antibiotics (2)
Brand
Sigma-Aldrich (2)
Sustainability
Biological Source
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Quality Segment
Application
Mode of Action
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
P8833
Puromycin dihydrochloride from
Streptomyces alboniger
,
powder, non-animal origin, suitable for cell culture, BioReagent
Expand
View Pricing
P7255
Puromycin dihydrochloride from
Streptomyces alboniger
,
≥98% (HPLC), powder
Expand
View Pricing