Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pur-A-Lyzer
™
Mega Dialysis Kit
Products
Product Category
kits (11)
Brand
Sigma-Aldrich (12)
Capacity (mL)
Length (cm)
Cap
Membrane Material
MWCO (kDa)
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
PURG10020
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 1000, capacity 3-20 mL, pkg of 5 Mega dialysis tubes (in individual pouch), MWCO 1 kDa, cap, 20 mL displacement
Expand
View Pricing
PURG12020
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 12000, capacity 3-20 mL, MWCO 12-14 kDa, cap, 20 mL displacement
Expand
View Pricing
PURG35020
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 3500, capacity 3-20 mL, MWCO 3.5 kDa, cap, 20 mL displacement
Expand
View Pricing
PURG10010
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 1000, capacity 3-20 mL, MWCO 1 kDa, cap, 10 mL displacement
Expand
View Pricing
PURG12010
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 12000, capacity 3-20 mL, MWCO 12-14 kDa, cap, 10 mL displacement
Expand
View Pricing
PURG10015
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 1000, capacity 3-20 mL, MWCO 1 kDa, cap, 15 mL displacement
Expand
View Pricing
PURG35015
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 3500, capacity 3-20 mL, MWCO 3.5 kDa, cap, 15 mL displacement
Expand
View Pricing
PURG35010
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 3500, capacity 3-20 mL, MWCO 3.5 kDa, cap, 10 mL displacement
Expand
View Pricing
PURG60010
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 6000, capacity 3-20 mL, MWCO 6-8 kDa, cap, 10 mL displacement
Expand
View Pricing
PURG12015
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 12000, capacity 3-20 mL, MWCO 12-14 kDa, cap, 15 mL displacement
Expand
View Pricing
PURG60020
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 6000, capacity 3-20 mL, MWCO 6-8 kDa, cap, 20 mL displacement
Expand
View Pricing
PURG60015
Pur-A-Lyzer
™
Mega Dialysis Kit,
Mega 6000, capacity 3-20 mL, MWCO 6-8 kDa, cap, 15 mL displacement
Expand
View Pricing