Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pt-Co/Hazen/APHA Colour Reference Standard
Products
Brand
Supelco (7)
Manufacturer
Physical Form
Agency
Format
Application
Reference Material Type
Special Grade
certified reference material (1)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
PTCO010
Pt-Co/Hazen/APHA Colour Reference Standard,
ASTM D 1209 (ASTM colour 10)
Expand
View Pricing
PTCO500
Pt-Co/Hazen/APHA Colour Reference Standard,
stock solution, ASTM D 1209 (ASTM colour 500)
Expand
View Pricing
PTCO050
Pt-Co/Hazen/APHA Colour Reference Standard,
ASTM D 1209 (ASTM colour 50)
Expand
View Pricing
PTCO100
Pt-Co/Hazen/APHA Colour Reference Standard,
ASTM D 1209 (ASTM colour 100)
Expand
View Pricing
PTCO015
Pt-Co/Hazen/APHA Colour Reference Standard,
ASTM D 1209 (ASTM colour 15)
Expand
View Pricing
PTCO005
Pt-Co/Hazen/APHA Colour Reference Standard,
ASTM D 1209 (ASTM colour 5)
Expand
View Pricing
PTCO030
Pt-Co/Hazen/APHA Colour Reference Standard,
ASTM D 1209 (ASTM colour 30)
Expand
View Pricing
PTCO000
Pt-Co/Hazen/APHA Colour Reference Standard,
ASTM D 1209 (ASTM colour 0)
Expand
View Pricing