
Propyl gallate for ID and assay

Synonym(s):
Propyl gallate, 3,4,5-Trihydroxybenzoic acid propyl ester
Linear Formula:
3,4,5-(HO)3C6H2CO2CH2CH2CH3
CAS Number:
Molecular Weight:
212.20
Products
Formula Weight
Manufacturer
Melting Point (°C)
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing