Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Propyl butyrate
Synonym(s):
n
-Propyl-
n
-butanoate
Linear Formula:
CH
3
CH
2
CH
2
COOCH
2
CH
2
CH
3
CAS Number:
105-66-8
Molecular Weight:
130.18
EC Number:
203-320-0
Products
Product Category
aroma chemicals (2)
building blocks (1)
Brand
Sigma-Aldrich (3)
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Halal (2)
Kosher (2)
Food Grade (1)
Natural (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
W293407
Propyl butyrate,
≥98%
Expand
View Pricing
291838
Propyl butyrate,
99%
Expand
View Pricing
W293415
Propyl butyrate,
natural, ≥95%, FG
Expand
View Pricing