Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ProClin
™
300
Products
Product Category
preservatives (6)
Brand
Sigma-Aldrich (6)
Color
pH Value
Physical Form
Chemical Composition
Quality Segment
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
48914-U
ProClin
™
300,
pkg of 400 mL, ProClin
™
preservative for use in IVD manufacturing, CMIT/MIT 3 %
Expand
View Pricing
48912-U
ProClin
™
300,
pkg of 50 mL, ProClin
™
preservative for use in IVD manufacturing, CMIT/MIT 3 %
Expand
View Pricing
48915-U
ProClin
™
300,
ProClin
™
preservative for use in IVD manufacturing, pkg of 2.0 L, CMIT/MIT 3 %
Expand
View Pricing
48918-U
ProClin
™
300,
pkg of 18 L, ProClin
™
preservative for use in IVD manufacturing, CMIT/MIT 3 %
Expand
View Pricing
48917-U
ProClin
™
300,
ProClin
™
preservative for use in IVD manufacturing, pkg of 3.6 L, CMIT/MIT 3 %
Expand
View Pricing
48911-U
ProClin
™
300,
ProClin
™
preservative for use in IVD manufacturing, CMIT/MIT 3%, Method Development Kit, pkg of 3 × 5 mL each (three different lots)
Expand
View Pricing