Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Precision Seal
®
rubber septa cap
Products
Product Category
septa (9)
Brand
Sigma-Aldrich (9)
Color
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z553891
Precision Seal
®
rubber septa cap,
white, for 5-6 mm O.D. tubes and ampules
Expand
View Pricing
Z554014
Precision Seal
®
rubber septa cap,
red, for 5-6 mm O.D. tubes and ampules
Expand
View Pricing
Z565768
Precision Seal
®
rubber septa cap,
white, for 7-8 mm O.D. tubes and ampules
Expand
View Pricing
Z764612
Precision Seal
®
rubber septa cap,
white, for 3 mm O.D. tubes and ampules, pack of 10 ea
Expand
View Pricing
Z565776
Precision Seal
®
rubber septa cap,
white, for 10 mm O.D. tubes and ampules
Expand
View Pricing
Z565792
Precision Seal
®
rubber septa cap,
red, for 10 mm O.D. tubes and ampules
Expand
View Pricing
Z764620
Precision Seal
®
rubber septa cap,
red, for 3 mm O.D. tubes and ampules
Expand
View Pricing
Z566543
Precision Seal
®
rubber septa cap,
white, for 9 mm O.D. tubes and ampules
Expand
View Pricing
Z565784
Precision Seal
®
rubber septa cap,
red, for 7-8 mm O.D. tubes and ampules
Expand
View Pricing