Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Precast Agarose Gels
Products
Brand
Sigma-Aldrich (5)
Physical Form
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
P6222
Precast Agarose Gels,
8-well (portrait orientation)
Expand
View Pricing
P5472
Precast Agarose Gels,
8-well (portrait orientation)
Expand
View Pricing
P5722
Precast Agarose Gels,
20-well (landscape orientation)
Expand
View Pricing
P6097
Precast Agarose Gels,
24 wells (2 × 12-well portrait orientation)
Expand
View Pricing
P5972
Precast Agarose Gels,
24 wells (2 × 12-well portrait orientation)
Expand
View Pricing