Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium thiocyanate
Synonym(s):
Potassium rhodanide, Potassium thiocyanate
Linear Formula:
KSCN
CAS Number:
333-20-0
Molecular Weight:
97.18
EC Number:
206-370-1
Products
Product Category
salts (5)
building blocks (1)
Brand
Sigma-Aldrich (2)
Supelco (2)
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
P3011
Potassium thiocyanate,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
207799
Potassium thiocyanate,
ACS reagent, 99%
Expand
View Pricing
1.05125
Potassium thiocyanate,
for analysis EMSURE
®
ACS,ISO,Reag. Ph Eur
Expand
View Pricing
1.05124
Potassium thiocyanate,
EMPLURA
®
Expand
View Pricing
P2713
Potassium thiocyanate,
BioXtra, ≥99.0%
Expand
View Pricing