Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium phosphate dibasic
Synonym(s):
Dipotassium hydrogenphosphate, Dipotassium phosphate, sec.-Potassium phosphate, Potassium phosphate dibasic
Linear Formula:
K
2
HPO
4
CAS Number:
7758-11-4
Molecular Weight:
174.18
EC Number:
231-834-5
Products
Product Category
buffers (5)
salts (5)
Brand
Sigma-Aldrich (7)
Supelco (4)
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Special Grade
Anhydrous (5)
ACS reagent (3)
Puriss (1)
Puriss P.A. (1)
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
P3786
Potassium phosphate dibasic,
ACS reagent, ≥98%
Expand
View Pricing
1.05104
Potassium phosphate dibasic,
anhydrous for analysis EMSURE
®
Expand
View Pricing
60356
Potassium phosphate dibasic,
puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Expand
View Pricing
60353
Potassium phosphate dibasic,
anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Expand
View Pricing
1.05109
Potassium phosphate dibasic,
anhydrous 99.99 Suprapur
®
Expand
View Pricing
P8281
Potassium phosphate dibasic,
reagent grade, ≥98.0%
Expand
View Pricing
5.43839
Potassium phosphate dibasic,
anhydrous for HPLC LiChropur
™
Expand
View Pricing
450200
Potassium phosphate dibasic,
99.95% trace metals basis
Expand
View Pricing
795496
Potassium phosphate dibasic,
anhydrous, free-flowing, Redi-Dri
™
, ACS reagent, ≥98%
Expand
View Pricing
P2222
Potassium phosphate dibasic,
meets USP testing specifications
Expand
View Pricing
59678
Potassium phosphate dibasic,
suitable for HPLC, LiChropur
™
, 99.0-101.0% (T)
Expand
View Pricing