Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium perchlorate
Synonym(s):
Potassium perchlorate
Linear Formula:
KClO
4
CAS Number:
7778-74-7
Molecular Weight:
138.55
EC Number:
231-912-9
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Special Grade
ACS reagent (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
241830
Potassium perchlorate,
ACS reagent, ≥99%
Expand
View Pricing
1.05076
Potassium perchlorate,
for analysis EMSURE
®
ACS
Expand
View Pricing
460494
Potassium perchlorate,
≥99.99% trace metals basis
Expand
View Pricing