Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium nitrate-
15
N
Synonym(s):
15
N Labeled KNO
3
Linear Formula:
K
15
NO
3
CAS Number:
57654-83-8
Molecular Weight:
102.10
Products
Product Category
salts (4)
stable isotopes (4)
Brand
Sigma-Aldrich (4)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Application
Markush Class
Markush Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
335134
Potassium nitrate-
15
N,
98 atom %
15
N
Expand
View Pricing
348481
Potassium nitrate-
15
N,
10 atom %
15
N
Expand
View Pricing
486124
Potassium nitrate-
15
N,
5 atom %
15
N
Expand
View Pricing
490547
Potassium nitrate-
15
N,
60 atom %
15
N
Expand
View Pricing