Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium hydroxide solution
Linear Formula:
KOH
CAS Number:
1310-58-3
Molecular Weight:
56.11
Products
Product Category
inorganic bases (2)
bases (1)
Brand
Sigma-Aldrich (1)
Supelco (10)
Formula Weight
Physical Form
Product Line
Agency
Quality Segment
Reaction Type
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
417661
Potassium hydroxide solution,
45 wt. % in H
2
O
Expand
View Pricing
P4494
Potassium hydroxide solution,
8.0 M KOH (8.0N), volumetric solution suitable for titration
Expand
View Pricing
1.09108
Potassium hydroxide solution,
c(KOH) = 1 mol/l (1 N), reag. USP, ready-to-use volumetric solution for titration, Titriplex
®
Expand
View Pricing
1.09112
Potassium hydroxide solution,
c(KOH) = 0.1 mol/l (0.1 N), reag. Ph. Eur., ready-to-use volumetric solution for titration, Titripur
®
Expand
View Pricing
1.60341
Potassium hydroxide solution,
c(KOH) = 0.5 mol/l (0.5 N), in ethanol, reag. USP, ready-to-use volumetric solution for titration in non-aqueous media, Titriplex
®
Expand
View Pricing
1.09107
Potassium hydroxide solution,
c(KOH) = 1 mol/l (1 N), max. 0.4 ppm Ca, ready-to-use volumetric solution for titration, Titriplex
®
Expand
View Pricing
1.09921
Potassium hydroxide solution,
c(KOH) = 0.1 mol/l (0.1 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol
®
Expand
View Pricing
1.09919
Potassium hydroxide solution,
c(KOH) = 0.5 mol/l (0.5 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol
®
Expand
View Pricing
67109
Potassium hydroxide solution,
4 M, eluent concentrate for IC
Expand
View Pricing
1.09918
Potassium hydroxide solution,
c(KOH) = 1 mol/l (1 N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol
®
Expand
View Pricing
1.11586
Potassium hydroxide solution,
c(KOH) = 0.5 mol/l (0.5 N), reag. USP, ready-to-use volumetric solution for titration, Titriplex
®
Expand
View Pricing