Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polymyxin B sulfate salt
Empirical Formula (Hill Notation):
C
55
H
96
N
16
O
13
· 2H
2
SO
4
CAS Number:
1405-20-5
Molecular Weight:
1385.61
EC Number:
215-774-7
Products
Product Category
antibiotics (2)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Physical Form
Product Line
Activity Spectrum
Quality Segment
Mode of Action
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
P4932
Polymyxin B sulfate salt,
powder, non-animal origin, suitable for cell culture, BioReagent
Expand
View Pricing
P1004
Polymyxin B sulfate salt,
Expand
View Pricing