Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly-ʟ-Lysine Hydrobromide
CAS Number:
25988-63-0
Products
Product Category
attachment factors (5)
Brand
Sigma-Aldrich (5)
Biological Source
Physical Form
Product Line
Quality Segment
Optical Isomer
Sterilization Method
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
P4707
Poly-ʟ-Lysine Hydrobromide,
synthetic, liquid, 0.01%, suitable for cell culture
Expand
View Pricing
P4832
Poly-ʟ-Lysine Hydrobromide,
synthetic, mol wt 150,000-300,000, solution, 0.01%, suitable for cell culture
Expand
View Pricing
P6282
Poly-ʟ-Lysine Hydrobromide,
synthetic, mol wt 70,000-150,000, powder, γ-irradiated, suitable for cell culture, BioXtra
Expand
View Pricing
P9155
Poly-ʟ-Lysine Hydrobromide,
synthetic, mol wt 30,000-70,000, powder, γ-irradiated, suitable for cell culture, BioXtra
Expand
View Pricing
P5899
Poly-ʟ-Lysine Hydrobromide,
synthetic, mol wt ≥300,000, powder, γ-irradiated, suitable for cell culture, BioXtra
Expand
View Pricing