Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(Glu, Tyr) sodium salt
CAS Number:
97105-00-5
Products
Product Category
amino acids (4)
Brand
Sigma-Aldrich (4)
Sustainability
Color
Feature
Physical Form
Quality Segment
Application
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P0275
Poly(Glu, Tyr) sodium salt,
Glu:Tyr (4:1), mol wt 20,000-50,000
Expand
View Pricing
P7244
Poly(Glu, Tyr) sodium salt,
Glu:Tyr (4:1), mol wt 5,000-20,000
Expand
View Pricing
81357
Poly(Glu, Tyr) sodium salt,
suitable for ligand binding assays, Mol wt 20,000-50,000
Expand
View Pricing
P0151
Poly(Glu, Tyr) sodium salt,
Glu:Tyr (1:1), mol wt 20,000-50,000
Expand
View Pricing