Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(ethylene glycol) bis(carboxymethyl) ether
Linear Formula:
HOOCCH
2
(OCH
2
CH
2
)
n
OCH
2
COOH
CAS Number:
39927-08-7
Products
Brand
Sigma-Aldrich (2)
Physical Form
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
407038
Poly(ethylene glycol) bis(carboxymethyl) ether,
average M
N
600, cross-linking reagent amine reactive, carboxylic acid
Expand
View Pricing
406996
Poly(ethylene glycol) bis(carboxymethyl) ether,
average M
N
250, cross-linking reagent amine reactive, carboxylic acid
Expand
View Pricing