Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly-D-lysine hydrobromide
Linear Formula:
D-Lys-(D-Lys)
n
-D-Lys · xHBr
CAS Number:
27964-99-4
Products
Product Category
amino acids (6)
Brand
Sigma-Aldrich (6)
Color
Physical Form
Functional Group
Optical Isomer
Amino Acid
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
P0899
Poly-D-lysine hydrobromide,
suitable for cell culture, Mol wt 70,000-150,000
Expand
View Pricing
P7886
Poly-D-lysine hydrobromide,
suitable for cell culture, Mol wt 30,000-70,000
Expand
View Pricing
P1149
Poly-D-lysine hydrobromide,
suitable for cell culture, Mol wt 150,000-300,000
Expand
View Pricing
P1024
Poly-D-lysine hydrobromide,
suitable for cell culture, Mol wt ≥300,000
Expand
View Pricing
P6403
Poly-D-lysine hydrobromide,
suitable for cell culture, Mol wt 4,000-15,000
Expand
View Pricing
P0296
Poly-D-lysine hydrobromide,
suitable for cell culture, Mol wt 1,000-5,000
Expand
View Pricing