Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly-DL-ornithine hydrobromide
CAS Number:
82682-33-5
Products
Product Category
amino acids (3)
Brand
Sigma-Aldrich (3)
Color
Physical Form
Quality Segment
Optical Isomer
Amino Acid
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
P8638
Poly-DL-ornithine hydrobromide,
suitable for cell culture, Mol wt 3,000-15,000
Expand
View Pricing
P0671
Poly-DL-ornithine hydrobromide,
suitable for cell culture, Mol wt >30,000
Expand
View Pricing
P0421
Poly-DL-ornithine hydrobromide,
mol wt 15,000-30,000
Expand
View Pricing