Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(acrylic acid)
Linear Formula:
(C
3
H
4
O
2
)
n
CAS Number:
9003-01-4
Products
Brand
Sigma-Aldrich (5)
Supelco (1)
Format
Quality Segment
Polymer Composition
Polymer Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
181285
Poly(acrylic acid),
average M
v
~450,000
Expand
View Pricing
323667
Poly(acrylic acid),
Expand
View Pricing
306215
Poly(acrylic acid),
average M
v
~1,250,000
Expand
View Pricing
306231
Poly(acrylic acid),
average M
v
~4,000,000
Expand
View Pricing
306223
Poly(acrylic acid),
average M
v
~3,000,000
Expand
View Pricing
181293
Poly(acrylic acid),
analytical standard, average M
n
130,000 (Typical)
Expand
View Pricing