Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum-rhodium alloy, Pt90Rh10
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 46
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
GF34062870
Platinum-rhodium alloy, Pt90Rh10,
0.1m wire, 1.0mm diameter, annealed
Expand
View Pricing
GF22099181
Platinum-rhodium alloy, Pt90Rh10,
1m wire, 0.005mm diameter, wollaston wire
Expand
View Pricing
GF33152341
Platinum-rhodium alloy, Pt90Rh10,
0.2m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF00827018
Platinum-rhodium alloy, Pt90Rh10,
0.2m wire, 0.005mm diameter, wollaston wire
Expand
View Pricing
GF74468605
Platinum-rhodium alloy, Pt90Rh10,
50mm tube, 0.94mm outside diameter, 0.12mm wall thickness, 0.7mm inside diameter, as drawn
Expand
View Pricing
GF05740961
Platinum-rhodium alloy, Pt90Rh10,
2m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF19942757
Platinum-rhodium alloy, Pt90Rh10,
foil, 25 x 25mm, 0.125mm thickness
Expand
View Pricing
GF67503663
Platinum-rhodium alloy, Pt90Rh10,
0.5m wire, 0.30mm diameter, annealed
Expand
View Pricing
GF85645929
Platinum-rhodium alloy, Pt90Rh10,
1m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF03376795
Platinum-rhodium alloy, Pt90Rh10,
0.5m wire, 0.005mm diameter, wollaston wire
Expand
View Pricing
GF19818861
Platinum-rhodium alloy, Pt90Rh10,
foil, 50 x 50mm, 0.125mm thickness
Expand
View Pricing
GF42805852
Platinum-rhodium alloy, Pt90Rh10,
100mm tube, 0.94mm outside diameter, 0.12mm wall thickness, 0.7mm inside diameter, as drawn
Expand
View Pricing
GF98706280
Platinum-rhodium alloy, Pt90Rh10,
0.2m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF25679391
Platinum-rhodium alloy, Pt90Rh10,
1m wire, 0.20mm diameter, annealed
Expand
View Pricing
GF65538085
Platinum-rhodium alloy, Pt90Rh10,
2m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF62397793
Platinum-rhodium alloy, Pt90Rh10,
0.5m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF03040513
Platinum-rhodium alloy, Pt90Rh10,
0.5m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF21624320
Platinum-rhodium alloy, Pt90Rh10,
0.1m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF24172994
Platinum-rhodium alloy, Pt90Rh10,
10m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF72212745
Platinum-rhodium alloy, Pt90Rh10,
2m wire, 0.05mm diameter, annealed
Expand
View Pricing
Page 1 of 3
Page 1 of 3